Merck
  • Detection of propan-1,2-diol in neonatal brain by in vivo proton magnetic resonance spectroscopy.
Detection of propan-1,2-diol in neonatal brain by in vivo proton magnetic resonance spectroscopy.
Magnetic resonance in medicine (1994-12-01)
E B Cady, A Lorek, J Penrice, E O Reynolds, R A Iles, S P Burns, G A Coutts, F M Cowan
ABSTRACT
Cerebral in vivo proton magnetic resonance spectroscopy of 13 newborn infants displaying seizures and receiving phenobarbitone, in one case supplemented by phenytoin, showed signals from propan-1,2-diol (the injection vehicle for both these anticonvulsants). Subsequent in vitro spectroscopy of cerebro spinal fluid (CSF) from one of these infants also showed signals from this substance. The estimated in vivo propan-1,2-diol concentration (approximately 3 mM) was less than that measured in the CSF sample (14.4 mM). These observations suggest that propan-1,2-diol may accumulate in cerebral tissue and misidentification of its signals in both in vivo and in vitro proton spectra may confuse diagnoses of metabolic or other disorders.
Product Description
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Glutamine
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
SAFC
L-Isoleucine
SAFC
L-Methionine
SAFC
Taurine
SAFC
L-Glutamine
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Adenosine 5′-diphosphate, ≥95% (HPLC)
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)