  • Search Results
  • Lower concentrations of B-vitamin subgroups in the serum and amniotic fluid correlate to cleft lip and palate appearance in the offspring of A/WySn mice.
Lower concentrations of B-vitamin subgroups in the serum and amniotic fluid correlate to cleft lip and palate appearance in the offspring of A/WySn mice.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2013-05-07)
Konstanze Scheller, Thomas Röckl, Christian Scheller, Johannes Schubert
ABSTRACT
The pathogenesis and prevention of cleft lip and palate (CL/P) have been studied mainly in clinical and animal experiments. A prophylactic poly-B-vitamin substitution during the first months of pregnancy has provided the most encouraging results for the prevention of CL/P recurrence in families at risk. In vitro studies of the palatal organ in an A/WySn mouse model have confirmed the positive influence of B-vitamins on palatal development. The present animal study was performed to analyze different B-vitamin concentrations in the serum and amniotic fluid of A/WySn mice according to the appearance of CL/P in their offspring. Concentrations of different B-vitamins (B1, B2, B3, B5, B6, and folic acid) in serum and amniotic fluid were analyzed by high-performance liquid chromatographic detection. Immunohistochemical staining against thiamin-1 receptor was performed on histologic midface sections of A/WySn fetuses with (n = 12) and without (n = 14) CL/P. Vitamin B5 (P < .001) and folic acid (P < .004) concentrations in the amniotic fluid of dams with CL/P were significantly lower than in dams without CL/P. Serum concentrations of folic acid (P = .5) and B5 (P = .4) showed no difference between the 2 groups. Dams with CL/P had significantly lower thiamine concentrations in serum (P = .01) and amniotic fluid (P < .001). Histologic midface sections presented high thiamin-1 receptor expression in the palatal shelf of fetuses with CL/P. A decreased use or uptake of some B-vitamin subgroups (B1, B5, and folic acid) in amniotic fluid and serum (vitamin B1) was correlated to an increased cleft appearance in A/WySn mice. The high thiamin-1 receptor expression in the palatal tissue of mouse fetuses with CL/P may be caused by a decreased availability of vitamin B1.
Product Description
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nitro Blue Tetrazolium, tablet
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Riboflavin, European Pharmacopoeia (EP) Reference Standard