Merck
  • From the Medical Board of the National Psoriasis Foundation: The risk of cardiovascular disease in individuals with psoriasis and the potential impact of current therapies.
From the Medical Board of the National Psoriasis Foundation: The risk of cardiovascular disease in individuals with psoriasis and the potential impact of current therapies.
Journal of the American Academy of Dermatology (2013-11-05)
Jeremy Hugh, Abby S Van Voorhees, Rajiv I Nijhawan, Jerry Bagel, Mark Lebwohl, Andrew Blauvelt, Sylvia Hsu, Jeffrey M Weinberg
ABSTRACT
Many studies have identified cardiovascular risk factors in patients with psoriasis. Some psoriasis therapies may increase cardiovascular disease (CVD) and others may decrease CVD. We reviewed the literature to define the impact of common psoriasis therapies on cardiovascular measures and outcomes. Phototherapy has no major cardiovascular impact and may reduce levels of proinflammatory cytokines. Acitretin increases serum lipids and triglycerides, but has not been shown to increase cardiovascular risk. Cyclosporine A increases blood pressure, serum triglycerides, and total cholesterol. Methotrexate is associated with a decreased risk of CVD morbidity and mortality. Among the biologics, data for tumor necrosis factor inhibitors suggest an overall reduction in cardiovascular events. Most data on short-term ustekinumab use suggest no effect on major adverse cardiovascular events, however some authorities remain concerned. Nevertheless, ustekinumab use over a 4-year period shows a decrease in major adverse cardiovascular events when compared both with the general US population and with psoriatics in Great Britain. Most studies lack the power and randomization of large clinical trials and long-term follow-up periods. In addition, the increased risk of CVD associated with psoriasis itself is a confounding factor. Some therapies for moderate to severe psoriasis, including methotrexate and tumor necrosis factor inhibitors, may reduce cardiovascular events in psoriatic patients. Ustekinumab appears to be neutral but there may be a long-term benefit. Appropriate patient counseling and selection and clinical follow-up are necessary to maximize safety with these agents. Further long-term study is necessary to quantify the benefits and risks associated with biologic therapies.
Product Description
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acitretin Related Compound A, United States Pharmacopeia (USP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Acitretin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyclosporin A solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Mycophenolic acid, ≥98%
Methotrexate, European Pharmacopoeia (EP) Reference Standard