Merck
  • Simultaneous determination of haloperidol and bromperidol and their reduced metabolites by liquid-liquid extraction and automated column-switching high-performance liquid chromatography.
Simultaneous determination of haloperidol and bromperidol and their reduced metabolites by liquid-liquid extraction and automated column-switching high-performance liquid chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-04-29)
Norio Yasui-Furukori, Yoshimasa Inoue, Misturu Chiba, Tomonori Tateishi
ABSTRACT
This study describes a new simultaneous determination of haloperidol and bromperidol and their reduced metabolites by modification of automated column-switching high-performance liquid chromatography. The test compounds were extracted from 1ml of plasma using chloroform-hexane (30:70 (v/v)), and the extract was injected into a hydrophilic metaacrylate polymer column for clean-up and a C(18) analytical column for separation. The mobile phases consisted of phosphate buffer (0.02M, pH 4.6), perchloric acid (60%) and acetonitrile (54:1:45 (v/v)) and was delivered at a flow-rate of 0.6ml/min. The peak was detected using a UV detector set at 215nm. The method was validated for the concentration range 1-100ng/ml, and good linearity (r >0.999) was confirmed. Intra-day coefficient variations (CVs) for haloperidol, reduced haloperidol, bromperidol and reduced bromperidol were less than 2.5, 3.1, 2.4 and 2.5%, respectively. Inter-day CVs for corresponding compounds were 3.9, 5.1, 2.6 and 4.4%, respectively. Relative errors ranged from -5 to 10% and mean recoveries were 96-100%. The limit of quantification was 1.0ng/m for each compound. This method shows good specificity with respect to commonly prescribed psychotropic drugs, and it could be successfully applied for pharmacokinetic studies and therapeutic drug monitoring, particularly in patients receiving both haloperidol and bromperidol.
Product Description
Bromperidol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.