Merck
  • NMDAR-regulated dynamics of layer 4 neuronal dendrites during thalamocortical reorganization in neonates.
NMDAR-regulated dynamics of layer 4 neuronal dendrites during thalamocortical reorganization in neonates.
Neuron (2014-04-02)
Hidenobu Mizuno, Wenshu Luo, Etsuko Tarusawa, Yoshikazu M Saito, Takuya Sato, Yumiko Yoshimura, Shigeyoshi Itohara, Takuji Iwasato
ABSTRACT
Thalamocortical (TC) connectivity is reorganized by thalamic inputs during postnatal development; however, the dynamic characteristics of TC reorganization and the underlying mechanisms remain unexplored. We addressed this question using dendritic refinement of layer 4 (L4) stellate neurons in mouse barrel cortex (barrel cells) as a model; dendritic refinement of L4 neurons is a critical component of TC reorganization through which postsynaptic L4 neurons acquire their dendritic orientation toward presynaptic TC axon termini. Simultaneous labeling of TC axons and individual barrel cell dendrites allowed in vivo time-lapse imaging of dendritic refinement in the neonatal cortex. The barrel cells reinforced the dendritic orientation toward TC axons by dynamically moving their branches. In N-methyl-D-aspartate receptor (NMDAR)-deficient barrel cells, this dendritic motility was enhanced, and the orientation bias was not reinforced. Our data suggest that L4 neurons have "fluctuating" dendrites during TC reorganization and that NMDARs cell autonomously regulate these dynamics to establish fine-tuned circuits.
Sigma-Aldrich
(S)-AMPA, ≥97%
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
(±)-AMPA, solid