Merck
The misleading appearance of silver nitrate on plain radiography.
Annals of the Royal College of Surgeons of England (2014-05-02)
N Narayan, M C Nogaro, A Littlewood, M D Latimer
ABSTRACT
Cauterisation with silver nitrate is commonly used to treat overgranulation. Silver nitrate has a high density and a high mass attenuation coefficient, and it is therefore highly radio-opaque. We present a case in which its topical application to an area of overgranulation was interpreted as a bony fragment by the reporting radiologist in a patient with a supracondylar humeral fracture whose radiograph after removal of K-wires showed a large radio-opaque lesion. Although not a new phenomenon, it is one not widely known, and it can lead to confusion and unnecessary further imaging.
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)