Merck
[Tin compounds in pharmacy and nutrition].
Acta pharmaceutica Hungarica (2001-02-24)
L Nagy, A Szorcsik, K Kovács
ABSTRACT
The occurrence of tin in plants, animals and humans is discussed, in relation to its abundance in the lithosphere and hydrosphere and the range of the different tin(II) and tin(IV) complexes formed. A reasoned consideration of its essentiality for living species is provided. It is concluded that tin is beneficial, even if not yet proved to be an essential element. After reference to the chemistry of tin compounds, there is a detailed discussion of their toxicity in animals and humans. Feasible routes for tin intake and uptake into humans are described. The past and current use of tin pharmaceuticals is reviewed and the areas for which they are currently permitted for use in humans as dentifrices and mouth washes, as radio-pharmaceuticals and for the treatment of jaundiced newborns are described. A detailed review of tin-containing antitumour agents as representative tin pharmaceuticals is also given. Finally, a list of tin-containing drugs and drug candidates is also shown.
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Tin, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.75%
Tin, foil, not light tested, 200x300mm, thickness 0.009mm, 97.4%
Tin, foil, not light tested, 150x150mm, thickness 0.02mm, 99.75%
Tin, wire reel, 2m, diameter 0.25mm, as drawn, 99.99+%
Tin, foil, not light tested, 50x50mm, thickness 0.015mm, 99.75%
Tin, foil, 8mm disks, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, not light tested, 25x25mm, thickness 0.004mm, 99.75%
Tin, microfoil, disks, 25mm, thinness 1.0μm, specific density 730μg/cm2, permanent mylar 3.5μm support, 99.999%
Tin, wire reel, 5m, diameter 1.5mm, as drawn, 99.99+%
Tin, wire reel, 0.2m, diameter 1.5mm, as drawn, 99.99+%
Tin, rod, 100mm, diameter 5.0mm, extruded, 99.75%
Tin, foil, not light tested, 25x25mm, thickness 0.01mm, 97.4%
Tin, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Tin, wire reel, 2m, diameter 1.5mm, as drawn, 99.99+%
Tin, rod, 100mm, diameter 6.0mm, 99.999+%
Tin, rod, 500mm, diameter 2.0mm, extruded, 99.75%
Tin, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Tin, rod, 1000mm, diameter 25.0mm, extruded, 99.75%
Tin, wire reel, 2m, diameter 0.5mm, as drawn, 99.99+%
Tin, foil, not light tested, 200x300mm, thickness 0.006mm, 97.4%
Tin, wire reel, 10m, diameter 1.0mm, as drawn, 99.99+%
Tin, foil, 8mm disks, thickness 0.006mm, 99.75%
Tin, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%