  • Search Results
  • Acute and chronic effects of botulinum neurotoxin a on the mammalian neuromuscular junction.
Acute and chronic effects of botulinum neurotoxin a on the mammalian neuromuscular junction.
Muscle & nerve (2013-11-13)
Padmamalini Baskaran, Baskaran Thyagarajan
ABSTRACT
Botulinum neurotoxin A (BoNT/A) cleaves SNAP-25 and inhibits acetylcholine (ACh) release at the neuromuscular junctions (NMJ) to cause neuroparalysis. Previous reports indicate a dyssynchrony between the inhibitory effect of BoNT/A on ACh release and SNAP-25 cleavage. We tested the in vitro (acute; 90 min) and in vivo (chronic; 12 h) effects of BoNT/A on stimulus-evoked ACh release (SEAR), twitch tension, and SNAP-25 cleavage in isolated extensor digitorum longus (EDL) nerve-muscle preparations (NMP). In vitro or in vivo BoNT/A poisoning inhibited SEAR and twitch tension. Conversely, SNAP-25 cleavage and inhibition of spontaneous release frequency were observed only in NMP poisoned with BoNT/A in vivo. Moreover, chronic treatment of BoNT/A inhibited ionomycin stimulated Ca(2+) signals in Neuro 2a cells. These results demonstrate that the inhibition of SEAR precedes SNAP-25 cleavage and suggest involvement of a more complex mechanism for the inhibitory effect of BoNT/A at the NMJ.
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Supelco
4-Aminopyridine, PESTANAL®, analytical standard
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
4-Aminopyridine, 98%
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
3,4-Diaminopyridine, ≥98%
Sigma-Aldrich
Acetylcholine perchlorate
Sigma-Aldrich
Ionomycin from Streptomyces conglobatus, ≥98% (HPLC)
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
USP
Acetylcholine chloride, United States Pharmacopeia (USP) Reference Standard
Acetylcholine chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri