  • Search Results
  • Combinational effects of farnesoid X receptor antagonist and statin on plasma lipid levels and low-density lipoprotein clearance in guinea pigs.
Combinational effects of farnesoid X receptor antagonist and statin on plasma lipid levels and low-density lipoprotein clearance in guinea pigs.
Life sciences (2014-05-09)
Yuichiro Amano, Eiichiro Ishikawa, Emiko Shinozawa, Mitsuyuki Shimada, Shotaro Miura, Ryutaro Adachi, Ryuichi Tozawa
ABSTRACT
We previously reported anti-dyslipidemic effects of a farnesoid X receptor antagonist in monkeys. In this study, we compared the cholesterol-lowering effects of single and combined administration of a farnesoid X receptor antagonist, compound-T8, and the 3-hydroxy-3-methylglutaryl-coenzyme A reductase inhibitor atorvastatin in a guinea pig model. Plasma levels of 7α-hydroxy-4-cholesten-3-one, a marker of hepatic cholesterol 7α-hydroxylase activity, were measured after a single administration of compound-T8. The effects of compound-T8 or atorvastatin on plasma cholesterol levels and low-density lipoprotein (LDL) clearance were investigated after 14 or 16 days of repeated dosing, respectively. Fractional catabolic rate of plasma LDL was estimated by intravenous injection of DiI-labeled human LDL. The cholesterol-lowering effects of combination therapy were investigated after 7 days of repeated treatment. Compound-T8 (10 and 30 mg/kg) increased plasma 7α-hydroxy-4-cholesten-3-one levels in a dose-dependent manner. Single administration of compound-T8 (30 mg/kg) and atorvastatin (30 mg/kg) reduced plasma non-high-density lipoprotein (non-HDL) cholesterol levels by 48% and 46%, respectively, and increased clearance of plasma DiI-labeled LDL by 29% and 35%, respectively. Compound-T8 (10mg/kg) or atorvastatin (10mg/kg) reduced non-HDL cholesterol levels by 19% and 25%, respectively, and combination therapy showed an additive effect and lowered cholesterol levels by 48%. Similar to atorvastatin, compound-T8 reduced plasma non-HDL cholesterol levels accompanied with accelerated LDL clearance in guinea pigs. Combination therapy additively decreased plasma non-HDL cholesterol levels. Therefore, monotherapy with a farnesoid X receptor antagonist and combination therapy of a farnesoid X receptor antagonist with atorvastatin would be attractive dyslipidemia treatment options.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%