  • Search Results
  • Dynamics of hippocampal acetylcholine release during lithium-pilocarpine-induced status epilepticus in rats.
Dynamics of hippocampal acetylcholine release during lithium-pilocarpine-induced status epilepticus in rats.
Journal of neurochemistry (2014-06-10)
Markus H Hillert, Imran Imran, Martina Zimmermann, Helene Lau, Stefanie Weinfurter, Jochen Klein
ABSTRACT
The lithium-pilocarpine model is a rat model of epilepsy that mimics status epilepticus in humans. Here, we report changes of acetylcholine (ACh) release in the hippocampus before, during and after status epilepticus as monitored by microdialysis in unanesthetized rats. Administration of pilocarpine (30 mg/kg s.c.) to rats pretreated with lithium chloride (127 mg/kg i.p.) caused a massive, six-fold increase of hippocampal ACh release, paralleling the development of tonic seizures. When seizures were stopped by administration of diazepam (10 mg/kg i.p.) or ketamine (75 mg/kg i.p.), ACh levels returned to normal. Extracellular concentrations of glutamate remained unchanged during this procedure. Administration of atropine (1 mg/kg i.p.) 2 h after pilocarpine caused a further increase of ACh but did not affect seizures, whereas injection of mecamylamine (5 mg/kg i.p.) reduced ACh levels and seizures in a delayed fashion. Local infusion of tetrodotoxin, 1 μM locally) or hemicholinium (10 μM locally) strongly reduced ACh release and had delayed effects on seizures. Administration of glucose or inositol (250 mg/kg each i.p.) had no visible consequences. In parallel experiments, lithium-pilocarpine-induced status epilepticus also enhanced striatal ACh release, and hippocampal ACh levels equally increased when status epilepticus was induced by kainate (30 mg/kg i.p.). Taken together, our results demonstrate that seizure development in status epilepticus models is accompanied by massive increases of extracellular ACh, but not glutamate, levels. Treatments that reduce seizure activity also reliably reduce extracellular ACh levels.
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Acetylcholine perchlorate
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
USP
Acetylcholine chloride, United States Pharmacopeia (USP) Reference Standard
Acetylcholine chloride, European Pharmacopoeia (EP) Reference Standard