Merck
  • Simplified extraction and cleanup for multiresidue determination of pesticides in lanolin.
Simplified extraction and cleanup for multiresidue determination of pesticides in lanolin.
Journal - Association of Official Analytical Chemists (1989-11-01)
H Diserens
ABSTRACT
In the proposed method, a light petroleum solution of lanolin (wool fat) is adsorbed on diatomaceous earth in an Extrelut column, and the pesticides are eluted with acetonitrile saturated with light petroleum. After evaporation to a small volume, the extract is subjected to solid-phase extraction (SPE) on a C-18 column. The acetonitrile eluate is evaporated to dryness and the residue is taken up in light petroleum. Organophosphorus pesticides are determined by temperature-programmed gas chromatography (GC) on a wide-bore column using a flame photometric detector in the phosphorus mode. Organochlorine pesticides are determined after miniaturized Florisil cleanup by classic GC on an OV-17/QF-1 packed column, using an electron capture detector. This procedure is more rapid and straightforward than the time-consuming AOAC extraction method, 29.014. Cleanup was better and the results obtained were comparable. Recoveries for 13 organochlorine and organophosphorus pesticides, frequently found in lanolin, ranged from 80 to 90%.
Sigma-Aldrich
Lanolin, tested according to Ph. Eur.
Pricing and availability is not currently available.