  • Search Results
  • Bleeding diathesis and gastro-duodenal ulcers in inherited cytosolic phospholipase-A2 alpha deficiency.
Bleeding diathesis and gastro-duodenal ulcers in inherited cytosolic phospholipase-A2 alpha deficiency.
Thrombosis and haemostasis (2014-08-12)
E M Faioni, C Razzari, A Zulueta, E A Femia, L Fenu, M Trinchera, G M Podda, M Pugliano, F Marongiu, M Cattaneo
ABSTRACT
Arachidonic acid (AA), when cleaved from phospholipids by cytosolic phospholipase A2 alpha (cPLA2a), generates eicosanoids, with pro-hemostatic, pro-inflammatory, vasoactive and gastro-protective functions. We describe a patient (27-year-old man) and his twin-sister with early-onset bleeding diathesis and recurrent gastro-intestinal (GI) ulcers. Platelet aggregation/δ-granules secretion by collagen was impaired, but normal by AA; serum levels of thromboxane (Tx) B2 and 12-hydroxyeicosatetraenoic acid, and urinary levels of 11-dehydro-TxB2 were extremely low. Patients were homozygous for 1723G>C transition in PLA2G4A gene, which changed the codon for Asp575 to His. GI ulcers affected 5/14 heterozygous (< 40 years) and 1/16 wild-type homozygous (> 60 years) family members; none had bleeding diathesis. The proband, his sister and mother also had mildly reduced factor XI levels. Platelet messenger RNA expression did not differ among subjects with different PLA2G4A genotypes. Conversely, platelet cPLA2a was undetectable by Western Blotting in the proband and his sister, and decreased in 1723G>C heterozygous subjects, suggesting that the variant is transcribed, but not translated or translated into an unstable protein. We described a syndromic form of deficiency of cPLA2a , characterised by recurrent GI ulcers and bleeding diathesis, associated with mild inherited deficiency of factor XI. Unlike other reported patients with cPLA2a deficiency, these patients had extremely low levels of platelet TxA2 biosynthesis.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)