  • Search Results
  • Comparative profiling of membrane lipids during water stress in Thellungiella salsuginea and its relative Arabidopsis thaliana.
Comparative profiling of membrane lipids during water stress in Thellungiella salsuginea and its relative Arabidopsis thaliana.
Phytochemistry (2014-10-14)
Buzhu Yu, Weiqi Li
ABSTRACT
The remodelling of membrane lipids contributes to the tolerance of plants to stresses, such as freezing and deprivation of phosphorus. However, whether and how this remodelling relates to tolerance of PEG-induced osmotic stress has seldom been reported. Thellungiella salsuginea is a popular extremophile model for studies of stress tolerance. In this study, it was demonstrated that T. salsuginea was more tolerant to PEG-induced osmotic stress than its close relative Arabidopsis thaliana. Lipidomic analysis indicated that plastidic lipids are more sensitive to PEG-induced osmotic stress than extra-plastidic ones in both species, and that the changes in plastidic lipids differed markedly between them. PEG-induced osmotic stress led to a dramatic decrease in levels of plastidic lipids in A. thaliana, whereas the change in plastidic lipid in T. salsuginea involved an adaptive remodelling shortly after the onset of PEG-induced osmotic stress. The two aspects of this remodelling involved increases in (1) the level of plastidic lipids, especially digalactosyl diacylglycerol, and (2) the double bond index of plastidic lipids. These remodelling steps could maintain the integrity and improve the fluidity of plastidic membranes and this may contribute to the PEG-induced osmotic stress tolerance of T. salsuginea.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
2′,7′-Dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Water, for TOC analysis
Supelco
Water, ACS reagent, for ultratrace analysis
SAFC
Sodium chloride solution, 5 M
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C