Merck
  • Repeated combined endovascular therapy with milrinone and nimodipine for the treatment of severe vasospasm: preliminary results.
Repeated combined endovascular therapy with milrinone and nimodipine for the treatment of severe vasospasm: preliminary results.
Acta neurochirurgica. Supplement (2014-11-05)
Camillo Sherif, Bernhard Wambacher, Michel Loyoddin, Radenko Karaic, Peter Krafft, Andreas Valentin, Dimiter Tscholakoff, Guenther Kleinpeter
ABSTRACT
Delayed vasospasm (VSP) following aneurysmal subarachnoid hemorrhage (aSAH) remains a major source of morbidity. Milrinone was recently suggested as an invasive VSP treatment option. It is a phosphodiesterase III inhibitor with vasodilating and additional positive inotrope and anti-inflammatory effects. In this preliminary series, we included patients with severe VSP and unsuccessful maximum conservative therapy. Inclusion criteria were (1) transcranial Doppler (TCD) mean >180 cm/s; (2) increase of >50 % of TCD mean values within 6 h to values >150 cm/s; and/or (3) neurological deterioration (after exclusion of hemorrhage, hydrocephalus, and other systemic reasons). Patients received endovascular therapy with nimodipine 2 mg followed by milrinone 4-8 mg. Reinterventions were indicated aggressively in cases of persistent neurological deficits or persistent high mean TCD >180 cm/s. Of 121 consecutive aSAH patients, 16 (13.2 %) received endovascular VSP therapy. Of these, 11 patients (68.5 %) received ≥ 3 interventions (median 4; maximum 9); 14 (87.5 %) showed postinterventional angiographic improvement of vessel diameters; and 11 (68.5 %) showed improvement of their neurological deficits after a mean follow-up time of 4.5 months. No cardiovascular adverse events attributed to milrinone were observed. Milrinone may be a useful supplementary substance for endovascular VSP therapy. Aggressive reintervention indications did not cause additional adverse events.
Sigma-Aldrich
Nimodipine
Sigma-Aldrich
Milrinone, ≥97% (TLC), powder
Nimodipine, European Pharmacopoeia (EP) Reference Standard
Supelco
Nimodipine, Pharmaceutical Secondary Standard; Certified Reference Material