Merck
  • Influence of organic carbon and metal oxide phases on sorption of 2,4,6-trichlorobenzoic acid under oxic and anoxic conditions.
Influence of organic carbon and metal oxide phases on sorption of 2,4,6-trichlorobenzoic acid under oxic and anoxic conditions.
Environmental monitoring and assessment (2014-12-01)
Isaac Ayodele Ololade, Nurudeen Abiola Oladoja, Folasade Alomaja, Oluwaranti Olubunmi Ololade, Esan O Olaseni, Femi Francis Oloye, Ruth O A Adelagun
ABSTRACT
Chlorobenzoic acids represent crucial recalcitrant metabolites in the environment; thus, the influence of soil components on the sorption of 2,4,6-trichlorobenzoic acid (TCB) under oxic and anoxic conditions was studied. The surficial physiognomies of untreated and isolated soil samples were studied using FTIR, XRD, specific surface area, and PZC determination. The roles of redox potential, dissolved organic carbon (DOC), and pH, particularly under anoxic condition, were appraised. Batch equilibrium adsorption studies on soils of variable Fe/Mn oxides and organic carbon showed that adsorption was low across all components (log Koc = 0.82-3.10 Lg(-1)). The sorption of 2,4,6-TCB was well described by the pseudo second-order kinetic model. The fluctuation of both redox potential and pH during anoxic experiment had a negative impact on the sorption, partitioning, and the oxidation of organic matter. Linear relationships were observed for Kd with both soil total organic carbon (TOC) and surface area (SA). The results showed the existence of DOC-mediated sorption of 2,4,6-TCB which seems to be enhanced at lower pH. The reductive dissolution, particularly of iron compounds, possibly impeded sorption of 2,4,6-TCB under anoxic condition. It could be inferred that habitats dominated by fluctuating oxygen concentrations are best suited for the development of environmental conditions capable of mineralizing 2,4,6-TCB and similar xenobiotics.
Product Description
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon, foil, 100x100mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 100x100mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon