  • Search Results
  • Impact of supramolecular interactions of dextran-β-cyclodextrin polymers on invertase activity in freeze-dried systems.
Impact of supramolecular interactions of dextran-β-cyclodextrin polymers on invertase activity in freeze-dried systems.
Biotechnology progress (2015-03-05)
Patricio R Santagapita, M Florencia Mazzobre, M Pilar Buera, Héctor L Ramirez, Leissy Gómez Brizuela, Horacio R Corti, Reynaldo Villalonga
ABSTRACT
β-Cyclodextrin (β-CD)-grafted dextrans with spacer arms of different length were employed to evaluate the impact of supramolecular interactions on invertase activity. The modified dextrans were used as single additives or combined with trehalose in freeze-dried formulations containing invertase. Enzyme activity conservation was analyzed after freeze-drying and thermal treatment. The change of glass transition temperature (Tg ) was also evaluated and related to effective interactions. Outstanding differences on enzyme stability were mainly related to the effect of the spacer arm length on polymer-enzyme interactions, since both the degree of substitution and the molecular weight were similar for the two polymers. This change of effective interactions was also manifested in the pronounced reduction of Tg values, and were related to the chemical modification of the backbone during oxidation, and to the attachment of the β-CD units with spacer arms of different length on dextran.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
D-Glucose-12C6, 99.5 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, ≥99%
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, ≥98.5% (HPLC)
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from starch, ≥99%