Merck
ISG12a mediates cell response to Newcastle disease viral infection.
Virology (2014-07-08)
Nianli Liu, Ying Long, Bin Liu, Darong Yang, Chen Li, Tianran Chen, Xiaohong Wang, Chen Liu, Haizhen Zhu
ABSTRACT
Newcastle disease virus (NDV) oncolysis is believed to be facilitated by a defective Type I interferon (IFN) response. We compared hepatocellular carcinoma (HCC)-derived cell lines and found that TRAIL-resistant cells were more susceptible to NDV oncolysis than were TRAIL-sensitive cells. In examining the IFN response, we found that basal expression of the IFN-stimulated gene (ISG)-12a was low in TRAIL-resistant but high in TRAIL-sensitive cells, and ISG12a over-expression or silencing enhanced or reduced their TRAIL sensitivities, respectively. Moreover, ISG12a over-expression in TRAIL-resistant cells decreased NDV replication but surprisingly increased oncolysis while ISG12a silencing had the opposite effect on TRAIL-sensitive cells. Finally, RIG-I and Noxa appear to also contribute to NDV oncolysis. Together, these results suggest that high basal ISG12a may inhibit NDV replication and oncolysis, while low basal ISG12a may allow sufficient NDV replication for induction of ISG12a, and other factors required for NDV oncolysis, with implications for future therapeutics.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sucrose, European Pharmacopoeia (EP) Reference Standard