  • Search Results
  • Chiral liquid chromatography-tandem mass spectrometry assay to determine that dexpramipexole is not converted to pramipexole in vivo after administered in humans.
Chiral liquid chromatography-tandem mass spectrometry assay to determine that dexpramipexole is not converted to pramipexole in vivo after administered in humans.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-10-08)
Dong Wei, Chichih Wu, Ping He, Doug Kerr, Scott Stecher, Liyu Yang
ABSTRACT
Dexpramipexole (DEX) was being investigated in clinical studies for the treatment of amyotrophic lateral sclerosis (ALS). To monitor the potential chiral interconversion of dexpramipexole to pramipexole (PPX) in vivo, a highly sensitive and selective chiral LC-MS/MS assay was developed and qualified for the detection of pramipexole in the presence of dexpramipexole in human plasma. In this assay, plasma samples were extracted by protein precipitation coupled with solid phase extraction (SPE). The analyte PPX was separated from its enantiomer DEX using a chiral HPLC method. The assay was qualified with a dynamic range of 0.150-1.00ng/mL. The lower limit of quantitation (LLOQ) for PPX was 0.150ng/mL in the presence of up to 1000ng/mL of DEX. The qualified method was used to analyze plasma samples from a DEX clinical study. No PPX was detected in humans at pharmacologically significant levels after administration of dexpramipexole at single doses up to 600mg per day.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, for residue analysis
Supelco
Ethanol, standard for GC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)