  • Search Results
  • Simultaneous determination of the repertoire of classical neurotransmitters released from embryonal carcinoma stem cells using online microdialysis coupled with hydrophilic interaction chromatography-tandem mass spectrometry.
Simultaneous determination of the repertoire of classical neurotransmitters released from embryonal carcinoma stem cells using online microdialysis coupled with hydrophilic interaction chromatography-tandem mass spectrometry.
Analytica chimica acta (2014-10-11)
Ya-Bin Tang, Fan Sun, Lin Teng, Wen-Bin Li, Shi-Min An, Chun Zhang, Xin-Jie Yang, Hao-Yu Lv, Xu-Ping Ding, Liang Zhu, Hong-Zhuan Chen
ABSTRACT
Dynamic, continuous, and simultaneous multi-analysis of transmitters is important for the delineation of the complex interactions between the neuronal and intercellular communications. But the analysis of the whole repertoire of classical transmitters of diverse structure is challenging due to their different physico-chemical properties and to their high polarity feature which leads to poor retention in traditional reversed-phase columns during LC-MS analysis. Here, an online microdialysis coupled with hydrophilic interaction chromatography-tandem mass spectrometry (online MD-HILIC-MS/MS) detection method was developed for the simultaneous measurement of the repertoire of classical transmitters (acetylcholine, serotonin, dopamine, norepinephrine, glutamate, GABA, and glycine). Stable isotope labeled internal standards and authentic matrix have been applied to guarantee reliable results. The method was successfully employed to reveal the characteristics of transmitter release from embryonal carcinoma stem cells. The method features simple procedure (no sample preparation), high recovery (≥ 73%), high accuracy (89.36%≤RE≤116.89%), good reproducibility (2.18%≤ RSD ≤14.56%), and sensitive limits of detection (2 pg for acetylcholine, serotonin, and glutamate, 10 pg for dopamine, norepinephrine, GABA, and glycine). It can be flexibly applied to determine the contents of the classical transmitters in other biological matrix samples with minor changes.
Product Description
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, certified reference material, TraceCERT®
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material