  • Search Results
  • Structural features of a novel polysaccharide isolated from a New Zealand Maori mushroom Iliodiction cibarium.
Structural features of a novel polysaccharide isolated from a New Zealand Maori mushroom Iliodiction cibarium.
Carbohydrate research (2015-02-07)
Lu Ren, Patrick J B Edwards, Conrad O Perera, Yacine Hemar
ABSTRACT
A purified water-soluble fraction (ICP5) of a polysaccharide, isolated from a local Maori mushroom Iliodiction cibarium in New Zealand, was investigated for its structural properties. Size exclusion chromatography and dynamic light scattering showed that ICP5 had a large MW of 1.6 × 10(5) Da with a hydrodynamic diameter of 83 ± 8 nm. Particle size measurements also displayed the tendency of ICP5 to aggregate when suspended in water. The results of GC-MS, FTIR and NMR analyses allowed some characteristics of the chemical structure of ICP5 to be determined. GC-MS results showed that ICP5 contained only glucose (81.61%), galactose (12.90%) and mannose (5.49%) monomers. The characterized fragment structures of ICP5 were found to be dominantly consisting of uronic acids, which formed a backbone containing 1,4-β-D-GlcpA. A small amount of unsaturated uronic acid also appeared to be present.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Hydrogen chloride, puriss., ≥99.8%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%