  • Search Results
  • Anti-inflammatory effect of Lycii radicis in LPS-stimulated RAW 264.7 macrophages.
Anti-inflammatory effect of Lycii radicis in LPS-stimulated RAW 264.7 macrophages.
The American journal of Chinese medicine (2014-07-10)
Mi Young Song, Hyo Won Jung, Seok Yong Kang, Kyung-Ho Kim, Yong-Ki Park
ABSTRACT
The root bark of Lycium barbarum (Lycii radicis cortex, LRC) is used as a cooling agent for fever and night sweats in East Asian traditional medicine. The inhibitory effect of LRC water extract on inflammation is unknown. In this study, the anti-inflammatory effect of LRC was investigated in lipopolysaccharide (LPS)-stimulated mouse macrophage, RAW 264.7 cells. LRC extract significantly decreased the LPS-induced production of inflammatory mediators, nitric oxide (NO), prostaglandin (PG) E2 and pro-inflammatory cytokines, interleukin (IL)-1β and IL-6 in the cells. In addition, LRC extract inhibited the LPS-induced expression of inducible NO synthase (iNOS) and cyclooxygenase (COX)-2 mRNA and protein, and inflammatory cytokines mRNA in the cells. The action mechanism of LRC underlies the blocking of LPS-mediated p38 and Jun N-terminal kinase (JNK), mitogen-activated protein kinases (MAPKs), and the nuclear factor (NF)-κB signaling pathway. These results indicate that LRC extract inhibits the inflammatory response in activated macrophages by down-regulating the transcription levels of inflammatory mediators and blocking the MAPKs and NF-κB pathway.
Product Description
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥99% (calc. to the dried substance)
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Monoclonal Anti-NOS2 antibody produced in mouse, clone 2G4, purified immunoglobulin, buffered aqueous solution
Sulfanilamide, European Pharmacopoeia (EP) Reference Standard
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Nitric Oxide Synthase, Inducible antibody produced in mouse, clone NOS-IN, ascites fluid
Dinoprostone, European Pharmacopoeia (EP) Reference Standard