  • Search Results
  • Multi-analyte high performance liquid chromatography coupled to high resolution tandem mass spectrometry method for control of pesticide residues, mycotoxins, and pyrrolizidine alkaloids.
Multi-analyte high performance liquid chromatography coupled to high resolution tandem mass spectrometry method for control of pesticide residues, mycotoxins, and pyrrolizidine alkaloids.
Analytica chimica acta (2015-03-04)
Zbynek Dzuman, Milena Zachariasova, Zdenka Veprikova, Michal Godula, Jana Hajslova
ABSTRACT
A new reliable and highly sensitive method based on high performance liquid chromatographic (HPLC) separation and high resolution tandem mass spectrometric detection (HRMS/MS) has been developed and validated for determination of 323 pesticide residues, 55 mycotoxins, and 11 plant toxins represented by pyrrolizidine alkaloids. The method was validated for three matrices, leek, wheat, and tea differing in nature/amount of co-extracts that may cause various matrix effects. For target analytes isolation, optimized QuEChERS-based (quick, easy, cheap, effective, rugged, and safe) extraction procedure was employed. Spectral HRMS/MS library has been established providing an entire spectrum of fragment ions for each analyte, which allows unbiased identification and confirmation of target compounds. The limits of quantification (LOQs) of target analytes were below 10 μg kg(-1) for 82%, 81%, and 61% for matrices leek, wheat, and tea, respectively. Recoveries were in the acceptable range (70-120%) according to SANCO/12571/2013 for most of target analytes, except for highly polar 'masked' mycotoxin deoxynivalenol-3-glucoside with recoveries 35%, 47%, and 42% for matrices leek, wheat, and tea, respectively. The linearities of calibration curves expressed as coefficients of determination were in the range of 0.9661-1.000, and repeatabilities expressed as relative standard deviations (RSDs) at LOQs lied in the range of 0.25-13.51%. The trueness of the method was verified using several certified reference materials (CRMs) and proficiency test samples.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%