  • Search Results
  • Liposome, gel and lipogelosome formulations containing sodium hyaluronate.
Liposome, gel and lipogelosome formulations containing sodium hyaluronate.
Journal of liposome research (2014-04-15)
Gülengül Duman, İsmail Aslan, A Yekta Özer, İbrahim İnanç, Alpay Taralp
ABSTRACT
The moisture-imparting effect of sodium hyaluronate (Na-HA) was investigated in liposome, gel and lipogelosome topical formulations. Sixteen liposome formulations were prepared with or without Na-HA (45 kDa) using various ratios of dimyristoylphosphatidylcholine, 1,2-dimyristoyl-sn-glycero-3-phosphatidylglycerol, dipalmitoylphosphatidylcholine and phospholipon 100H. The liposomes were characterized in terms of their structure, composition, zeta potential, Na-HA-entrapment capacity and stability. In particular, scanning electron microscopy, polarized light microscopy, dynamic light scattering and atomic force microscopy were utilized to probe appearance, size and size distribution and lamellarity. The work was then extended to gels using the gelling agents poloxamer (PXM 188 or 407) and Carbopol or Ultrez 21 (U-21), yielding liposome-loaded gel formulations (i.e. lipogelosomes). The in vitro release kinetics of Na-HA from liposomes, lipogelosomes and commercial Na-HA reference formulations were studied via a flow-through cell method. Among the liposomal formulations tested, L6, comprising of Na-HA-loaded phospholipon 100H:stearylamine:cholesterol (7:1:2), displayed optimal traits. The mean particle size, zeta potential and entrapment capacity of L6 were determined as 1900 nm, -20.9 mV and 15.0%. The optimum lipogelosome, LG4, was obtained by incorporating liposome L6 into a U-21 gel at a ratio of 1:1 (w/w). In clinical trials, in-house formulations were applied twice daily to 15 female volunteers. The two-week benefits were assessed against a commercial product; and in all cases, changes of skin humidity, sebum content, pH and wrinkle depth were promising. In particular, the LG4 lipogelosome-based formulation had significantly improved skin hydration and compliance, as evidenced by a moisture content gain of 30.4%.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dihexadecyl phosphate
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer