  • Search Results
  • Gonadotropin regulation and role of ovarian osteopontin in the periovulatory period.
Gonadotropin regulation and role of ovarian osteopontin in the periovulatory period.
The Journal of endocrinology (2014-10-30)
Yoshimitsu Kuwabara, Akira Katayama, Ryoko Tomiyama, Hu Piao, Sachiko Kurihara, Shuichi Ono, Katsuya Mine, Shigeo Akira, Hideo Orimo, Toshiyuki Takeshita
ABSTRACT
Osteopontin (OPN), a secreted glycoprotein, has multiple physiological functions. This study investigated the regulation and roles of OPN in the mouse ovary during the periovulatory stages. Immature female mice were treated with pregnant mare serum gonadotropin (PMSG) and human chorionic gonadotropin (hCG) to simulate follicle maturation and ovulation. In situ hybridization and real-time RT-PCR were performed to assess expression of Opn in the periovulatory ovary. Granulosa cells (GCs) from PMSG-primed immature mice were cultured with or without hCG in the presence or absence of OPN, and effects on expression of Opn, progesterone synthesis, and vascular endothelial growth factor (VEGF) signaling were assessed by real-time RT-PCR, ELISA, and western blotting analysis. Opn transcripts were significantly upregulated 3 h after hCG treatment, followed by a peak at 16 h, and the transcripts localized to GCs. Incubation with hCG significantly increased quantities of Opn transcripts in GCs and OPN levels in the culture medium at 12 and 24 h. Furthermore, OPN treatment caused a significant increase in the levels of Star protein, P 450 cholesterol side-chain cleavage enzyme (p450scc), 3-beta-hydroxysteroid dehydrogenase (Hsd3b), and progesterone in the culture medium. OPN treatment promoted Vegf expression in GCs, which was significantly suppressed by a phosphoinositide 3-kinase (PI3K) inhibitor. In addition, OPN treatment stimulated phosphorylation of AKT, a downstream PI3K signaling molecule. In conclusion, expression of Opn was upregulated in mouse ovarian GCs in response to a gonadotropin surge through epidermal growth factor receptor signaling, which enhances progesterone synthesis and Vegf expression during the early-luteal phase.
Sigma-Aldrich
p-Toluidine, 99%
Supelco
p-Toluidine, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Sigma-Aldrich
p-Toluidine, 99.6%
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Digoxigenin, European Pharmacopoeia (EP) Reference Standard