  • Search Results
  • Novel extracellular matrix biomarkers as predictors of adverse outcome in chronic heart failure: association between biglycan and response to statin therapy in the CORONA trial.
Novel extracellular matrix biomarkers as predictors of adverse outcome in chronic heart failure: association between biglycan and response to statin therapy in the CORONA trial.
Journal of cardiac failure (2014-12-03)
Thor Ueland, Pål Aukrust, Ståle H Nymo, John Kjekshus, John J V McMurray, John Wikstrand, Dirk Block, Christian Zaugg, Lars Gullestad
ABSTRACT
The extracellular matrix (ECM) plays an important role in left ventricular remodeling and progression of heart failure (HF). Biglycan and mimecan are ECM proteins that are abundantly expressed in cardiac tissue but have not been evaluated as prognostic markers in HF. We investigated their interaction with statin treatment and association with adverse outcome in chronic HF. The association between serum levels of biglycan and mimecan and the primary end point (cardiovascular [CV] death, nonfatal myocardial infarction, nonfatal stroke), all-cause mortality, CV death, the composite of all-cause mortality/hospitalization for worsening of HF, and the coronary end point was evaluated in 1,390 patients >60 years of age with ischemic systolic HF in the Controlled Rosuvastatin Multinational Trial in HF (CORONA) population, randomly assigned to 10 mg rosuvastatin or placebo. Serum biglycan and mimecan added no prognostic information beyond conventional risk factors, including N-terminal pro-B-type natriuretic peptide. However, statin treatment improved all outcomes except CV death in patients with low biglycan levels (ie, lower tertile), even after full multivariable adjustment. Although circulating levels of mimecan and biglycan were of limited predictive value in patients with chronic HF, circulating biglycan could be a useful marker for targeting statin therapy in patients with HF.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥40%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrofluoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥48%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)