  • Search Results
  • Size and concentration determination of (functionalised) fullerenes in surface and sewage water matrices using field flow fractionation coupled to an online accurate mass spectrometer: method development and validation.
Size and concentration determination of (functionalised) fullerenes in surface and sewage water matrices using field flow fractionation coupled to an online accurate mass spectrometer: method development and validation.
Analytica chimica acta (2015-04-08)
Pol Herrero, Patrick S Bäuerlein, Erik Emke, Rosa M Marcé, Pim de Voogt
ABSTRACT
In order to assess the environmental risks of a compound it is imperative to have suitable and reliable techniques for its determination in environmental matrices. In this paper, we focused on a method development for the recently introduced online coupling of a field flow fractionation (FFF) system to an Orbitrap-HRMS, that allows the simultaneous size and concentration determination of different aqueous fullerene aggregates and their concentrations in different size fractions. A 0.05% NH4OH solution in water was identified as the best carrier liquid for the analysis of the three different aqueous fullerene suspensions (C60 [60], [6,6]-phenyl-C61 butyric acid methyl ester ([60]PCBM) and [6,6]-(bis)phenyl-C61 butyric acid methyl ester ([60]bisPCBM)). The multi-angle light scattering (MALS) data received after employing the ammonia solution was consistent with both the theory and calibration using well defined Au and latex particles. The LODs obtained using Orbitrap HRMS detection were 0.1 μg L(-1) for an injection volume of 100 μL which are significantly better than the LODs obtained by using UV (20 μg L(-1)) and MALS detectors (5 μg L(-1)). However, these LODs can be further improved as in theory there is no limit to the amount of sample that can be injected into the FFF. Environmental samples (river and sewage water) were spiked with fullerenes and the fractograms obtained for these samples revealed that the matrix does affect the size of fullerene aggregates. Information on the size distribution can be useful for the risk assessment of these particles.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
SAFC
Sodium chloride solution, 5 M
Supelco
Toluene, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl