Merck
  • Electrochemiluminescence immunosensor using poly(l-histidine)-protected glucose dehydrogenase on Pt/Au bimetallic nanoparticles to generate an in situ co-reactant.
Electrochemiluminescence immunosensor using poly(l-histidine)-protected glucose dehydrogenase on Pt/Au bimetallic nanoparticles to generate an in situ co-reactant.
The Analyst (2014-06-19)
Lijuan Xiao, Yaqin Chai, Haijun Wang, Ruo Yuan
ABSTRACT
In this work, Pt/Au bimetallic nanoparticles (Pt/Au NPs) were used as nanocarriers to develop an electrochemiluminescence (ECL) immunosensor for sensitive cardiac troponin I (cTnI) detection, coupling with enzyme-based signal amplification. First, gold nanoparticles modified Ru(phen)3(2+)-doped silica nanoparticles (Au@RuSiO2 NPs) with numerous luminophores were used as a platform, potentially increasing the signal intensity. Second, Pt/Au NPs with large surface area and rich surface atoms were a superior matrix for the immobilization of numerous antibodies (Ab2), poly(l-histidine) (PLH) and glucose dehydrogenase (GDH). More importantly, the PLH-protected GDH exhibited excellent enzymatic activity for the oxidation of glucose accompanied by the reduction of NAD(+) to NADH. The in situ generated NADH acted as a co-reactant of Ru(phen)3(2+), significantly enhancing the ECL signal. In this manner, the designed immunosensor displayed high sensitivity for the detection of cTnI in the range of 0.010 ng mL(-1) to 10 ng mL(-1) with a detection limit of 3.3 pg mL(-1) (S/N = 3). The proposed strategy holds a new promise for highly sensitive bioassays for application in clinical analyses.
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)