  • Search Results
  • Curcumin induces apoptosis in breast cancer cells and inhibits tumor growth in vitro and in vivo.
Curcumin induces apoptosis in breast cancer cells and inhibits tumor growth in vitro and in vivo.
International journal of clinical and experimental pathology (2014-07-18)
Zhi-Dong Lv, Xiang-Ping Liu, Wei-Jun Zhao, Qian Dong, Fu-Nian Li, Hai-Bo Wang, Bin Kong
ABSTRACT
Curcumin has shown therapeutic and/or adjuvant therapeutic effects on the treatment of some patients with breast cancer. However, its mechanisms of action are largely unknown. This study was designed to investigate its antitumor effect and underlying mechanisms in human breast cancer MDA-MB-231 and MCF-7 cells. The MTT assay was used to evaluate cell viability, and flow cytometry, acridine orange staining and transmission electron microscopy were used to detect apoptosis for cultured cells. The protein expression in cells was evaluated by western blot analysis. Breast tumors were established by subcutaneous injection of MDA-MB-231 cells in nude BALB/c mice, and curcumin was administered to the mice. The size of tumors was monitored and the weight of tumors was examined. The exposure of breast cancer cells to curcumin resulted in growth inhibition and the induction of apoptosis in a dose-dependent manner. We also found that the expression of Bcl-2 protein decreased and the expression of Bax protein increased which lead to an increase of the Bax/Bcl-2 ratio. In mice bearing MDA-MB-231 xenograft tumors, administration of curcumin showed a significant decrease of tumor volumes and tumor weight compared with the control. Our results showed that curcumin exhibited antitumor effects in breast cancer cells with an induction of apoptosis.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O
Supelco
Dimethyl sulfoxide, analytical standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Curcumin, primary reference standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)