  • Search Results
  • Efficient extraction of bagasse hemicelluloses and characterization of solid remainder.
Efficient extraction of bagasse hemicelluloses and characterization of solid remainder.
Bioresource technology (2015-03-10)
Shuangquan Yao, Shuangxi Nie, Yue Yuan, Shuangfei Wang, Chengrong Qin
ABSTRACT
To reduce the degradation of cellulose and obtain high molecular weight of hemicellulose from the extracts, pH pre-corrected hot water pretreatment was developed by employing sodium hydroxide (3.9mol/L). The response surface model was established to optimize the extraction process. The species composition and purity of hemicellulose extract was analyzed by High Performance Liquid Chromatography (HPLC). The obtained solid remainder was analyzed by FTIR and SEM. The results showed that the component of xylose in hemicellulose extract was similar with commercial xylan. FTIR and SEM were shown to be able to evaluate solid remainder composition and surface characterization of the bagasse. The biggest balance between solid remainder and dissolved solid was obtained. Not only the yield of dissolved solid was improved, but the structure of solid remainder was also proved, which was beneficial to pulping and papermaking.
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Xylose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Galactose, plant-derived
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
D-(−)-Arabinose, ≥98%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
Sigma-Aldrich
D-(+)-Galactose, ≥98%
Sigma-Aldrich
D-(+)-Galactose, ≥99%
Sigma-Aldrich
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99%