  • Search Results
  • Development of a broad spectrum method for measuring flame retardants - overcoming the challenges of non-invasive human biomonitoring studies.
Development of a broad spectrum method for measuring flame retardants - overcoming the challenges of non-invasive human biomonitoring studies.
Analytical and bioanalytical chemistry (2014-08-31)
Agnieszka Kucharska, Adrian Covaci, Guido Vanermen, Stefan Voorspoels
ABSTRACT
Flame retardants (FRs), such as polybrominated diphenyl ethers (PBDEs) and phosphate flame retardants (PFRs), are a diverse group of compounds that are used to improve fire safety in many consumer products, such as furniture, textiles, electronics, etc. As these compounds are potentially harmful for human health, there is a need to better understand human exposure. Exposure to environmental contaminants can be monitored by the measurement of external sources of exposures and also by the determination of contaminant levels in human samples. For ethical and practical reasons, noninvasive matrices, such as hair, are preferred but, unfortunately, not widely used due to methodological limitations. A major challenge is sample availability: only small amounts can be sampled per individual. Multi-residue methods are therefore essential in order to determine multiple compounds in low sample amounts. In the framework of the FP7 project (INFLAME), an analytical method for the simultaneous determination of PBDEs and PFRs in human hair has been optimized and validated. Before extraction, hair samples (200 mg) were denaturated in nitric acid (HNO3) for 25 min at 25 °C. Consecutively, the samples were extracted using a mixture of hexane:dichloromethane, and extracts were further fractionated on Florisil. Fraction A which contained PBDEs was additionally cleaned on acidified silica gel and measured by gas chromatography coupled with electron capture negative ionization mass spectrometry (GC-ECNI-MS), while fraction B containing PFRs was directly analyzed by liquid chromatography tandem mass spectrometry (LC-MS/MS). This approach resulted in recoveries between 81-120% for PBDEs and 75-113% for PFRs (relative standard deviation (RSD) < 16%, n = 9). The optimized multi-residue method has been applied to 20 human hair samples. The obtained results indicated that the levels of PBDEs in hair samples were very low (0.2-12 ng/g) in relation to PBDE levels in human hair samples from other studies and most of the time below the method limit of quantification (LOQm). On the contrary, the PFR levels were relatively high as they were in the range of the levels previously found in dust samples (2-5,032 ng/g hair). We would like to highlight that the contribution of air and dust cannot be neglected (especially in the case of PFRs); therefore, we suggest that hair might be a good indicator of retrospective and integral exposure (which includes atmospheric deposition as well as endogenous mechanisms). Moreover, the aim of our study is focused on exposure assessment and levels detected in hair (independently of whether they come from internal or external exposure) and will significantly contribute to the exposure assessment.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%