  • Search Results
  • Determinants of intestinal permeability in healthy first-degree relatives of individuals with Crohn's disease.
Determinants of intestinal permeability in healthy first-degree relatives of individuals with Crohn's disease.
Inflammatory bowel diseases (2015-03-04)
David Kevans, Williams Turpin, Karen Madsen, Jon Meddings, Konstantin Shestopaloff, Wei Xu, Gabriel Moreno-Hagelsieb, Anne Griffiths, Mark S Silverberg, Andrew Paterson, Kenneth Croitoru
ABSTRACT
The Genetics, Environmental, Microbial Project is a multicenter study assessing etiological factors in Crohn's disease by studying healthy first-degree relatives (FDRs) of individuals affected by Crohn's disease. We aimed to evaluate the contribution of genetic, microbial, and environmental factors to the determination of intestinal permeability in healthy FDRs. IP was assessed using the lactulose-mannitol ratio (LacMan ratio). FDRs were genotyped for 167 inflammatory bowel disease-associated single nucleotide polymorphisms. Taxonomic profile of the fecal microbiota was determined by Illumina MiSeq pyrosequencing of 16S ribosomal RNA. The associations of LacMan ratio with demographic factors, inflammatory bowel disease-associated single nucleotide polymorphisms and the fecal microbiota were assessed. One thousand, one hundred ninety-six white FDRs were included [corrected]. Eleven percent of FDRs had an elevated LacMan ratio (≥0.03). A multivariate analysis demonstrated that younger subjects and nonsmokers had higher LacMan ratios, P = 3.62 × 10⁻⁴ and P = 0.03, respectively. The LacMan ratio was not significantly heritable, H2r, 0.13, P = 0.13. There was no association between any of the 167 inflammatory bowel disease-associated risk variants and LacMan ratio nor was there a correlation between fecal microbial composition and the LacMan ratio. We did not find LacMan ratio to be significantly heritable suggesting that the contribution of genetic factors to the determination of intestinal permeability in healthy FDRs is modest. Environmental factors, such as smoking, are likely more important determinants. The effect of age on intestinal barrier function has been underappreciated.
Sigma-Aldrich
Lactulose, ≥95%
Sigma-Aldrich
Lactulose, ≥98.0% (HPLC)
Lactulose, European Pharmacopoeia (EP) Reference Standard
USP
Lactulose, United States Pharmacopeia (USP) Reference Standard
Supelco
Lactulose, Pharmaceutical Secondary Standard; Certified Reference Material
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.