  • Search Results
  • Effect of commonly used organic solvents on aldehyde oxidase-mediated vanillin, phthalazine and methotrexate oxidation in human, rat and mouse liver subcellular fractions.
Effect of commonly used organic solvents on aldehyde oxidase-mediated vanillin, phthalazine and methotrexate oxidation in human, rat and mouse liver subcellular fractions.
Xenobiotica; the fate of foreign compounds in biological systems (2014-02-19)
Dayanidhi Behera, Rambabu Pattem, Girish Gudi
ABSTRACT
1. Aldehyde oxidase (AOX) is a cytosolic molybdoflavoprotein enzyme widely distributed across many tissues. In this study, we report the effect of commonly used organic solvents such as dimethyl sulfoxide (DMSO), acetonitrile (ACN), methanol and ethanol on AOX activity in human, rat and mouse liver S9 fractions using vanillin, phthalazine and methotrexate as probe substrates. 2. Methanol was found to be the most potent solvent in inhibiting vanillic acid and 1-phthalazinone formation in comparison to DMSO, ACN and ethanol across the species tested, except 7-hydroxy methotrexate. 3. Treatment with these solvents at approximate IC50 (% v/v) concentrations showed significant reduction in Clint and Vmax of the probe substrates and also resulted in different effects on Km across the species. 4. Marked differences in the activity and affinity towards AOX were observed with different probe substrates with methotrexate showing least activity and affinity as compared to vanillin and phthalazine. 5. Overall, AOX activity seemed to be more resilient to the presence of organic solvents at higher concentrations in human and rodent species. These results suggest that low concentrations of organic solvents are acceptable for in vitro incubations involving AOX-mediated metabolism.
Product Description
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Vanillin, certified reference material, TraceCERT®
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Vanillic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
USP
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%