  • Search Results
  • The effects of curcumin on depressive-like behavior in mice after lipopolysaccharide administration.
The effects of curcumin on depressive-like behavior in mice after lipopolysaccharide administration.
Behavioural brain research (2014-08-19)
Zhen Wang, Qingrui Zhang, Lin Yuan, Shuanglian Wang, Liwei Liu, Xudong Yang, Gang Li, Dexiang Liu
ABSTRACT
Current evidence supports that inflammation and increased cytokine levels are associated with depression-like symptoms and neuropsychological disturbances in humans. Curcumin has anti-inflammatory, antioxidant and anti-depressant-like properties. Here, we examined the effects of curcumin on lipopolysaccharide (LPS)-induced depressive-like behavior and inflammation in male mice. A single administration of LPS (0.83mg/kg, i.p.) increased the immobility time in the forced swimming test (FST) and tail suspension test (TST), reduced sucrose consumption without affecting spontaneous locomotor activity. Pretreatment with curcumin (50mg/kg, i.p.) for 7 consecutive days reversed LPS-induced alterations in the FST, TST, and sucrose preference test. Moreover, pre-treatment with curcumin attenuated LPS-induced microglial activation and overproduction of pro-inflammatory cytokine (interleukin-1β and tumor necrosis factor-α), as well as the levels of inducible nitric oxide synthase and cyclooxygenase-2 mRNA in the hippocampus and prefrontal cortex (PFC). In addition, curcumin ameliorated LPS-induced NF-κB activation in the hippocampus and PFC. The results demonstrate that curcumin may be an effective therapeutic agent for LPS-induced depressive-like behavior, partially due to its anti-inflammatory aptitude.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Curcumin, primary reference standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)