  • Search Results
  • Encapsulating gold nanomaterials into size-controlled human serum albumin nanoparticles for cancer therapy platforms.
Encapsulating gold nanomaterials into size-controlled human serum albumin nanoparticles for cancer therapy platforms.
Journal of microencapsulation (2014-08-05)
Donna V Peralta, Jibao He, Damon A Wheeler, Jin Z Zhang, Matthew A Tarr
ABSTRACT
Progress has been made in using human serum albumin nanoparticles (HSAPs) as promising colloidal carrier systems for early detection and targeted treatment of cancer and other diseases. Despite this success, there is a current lack of multi-functional HSAP hybrids that offer combinational therapies. The size of the HSAPs has crucial importance on drug loading and in vivo performance and has previously been controlled via manipulation of pH and cross-linking parameters. Gold nanomaterials have also gained attention for medicinal use due to their ability to absorb near-infrared light, thus offering photothermal capabilities. In this study, the desolvation and cross-linking approach was employed to encapsulate gold nanorods, nanoparticles, and nanoshells into HSAPs. Incorporation of gold nanomaterials caused some changes in HSAP sizes, but the general size trends remained. This encasement strategy facilitated size-controlled HSAPs, in the range of 100-300 nm, loaded with gold nanostructures; providing composite particles which incorporate photothermally active components.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis