  • Search Results
  • Formation of 3-monochloro-1,2-propanediol (3-MCPD) di- and monoesters from tristearoylglycerol (TSG) and the potential catalytic effect of Fe²⁺ and Fe³⁺.
Formation of 3-monochloro-1,2-propanediol (3-MCPD) di- and monoesters from tristearoylglycerol (TSG) and the potential catalytic effect of Fe²⁺ and Fe³⁺.
Journal of agricultural and food chemistry (2015-01-31)
Zhongfei Zhang, Boyan Gao, Xiaowei Zhang, Yuanrong Jiang, Xuebing Xu, Liangli Lucy Yu
ABSTRACT
This study investigated whether and how triacylglycerol (TAG) may serve as a precursor for 3-monochloro-1,2-propanediol (3-MCPD) fatty acid ester formation using tristearoylglycerol (TSG). TSG was reacted with inorganic chloride compounds including NaCl, KCl, FeCl2, CuCl2, ZnCl2, FeCl3 and dry HCl, or organic chlorine compound lindane at different temperatures. Only FeCl2 and FeCl3 were able to form 3-MCPD esters from TSG. Further electron spin resonance (ESR) determination of TSG, Fe2(SO4)3 and 5,5-dimethylpyrroline-N-oxide (DMPO) reactions revealed potential of Fe ion in promoting free radical generations under the experimental conditions. To further confirm the effect of Fe ion, chelating agent (EDTA-2Na) was added to the model reactions. The results showed for the first time that EDTA-2Na was able to reduce the generation of 3-MCPD esters. In addition, FT-IR examination indicated a possible involvement of a carbonyl group during the reaction. Taking all the observations together, the possible mechanisms, involving the formation of either a cyclic acyloxonium or a glycidol ester radical intermediate, were proposed for generating 3-MCPD fatty acid di- and mono- esters from TAG under a high temperature and low moisture condition, as well as the coformation of glycidol esters. The results from this study may be useful for reducing the level of 3-MCPD esters and related toxicants in the refined edible oils and food products.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Toluene, analytical standard
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Supelco
5,5-Dimethyl-1-pyrroline N-oxide, for ESR-spectroscopy
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous