  • Search Results
  • A liquid chromatography-tandem mass spectrometry assay for quantification of rilpivirine and dolutegravir in human plasma.
A liquid chromatography-tandem mass spectrometry assay for quantification of rilpivirine and dolutegravir in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-28)
M Grégoire, G Deslandes, C Renaud, R Bouquié, C Allavena, F Raffi, P Jolliet, E Dailly
ABSTRACT
A liquid chromatography-tandem mass spectrometry assay requiring a 100μL aliquot of human plasma for simultaneous determination of rilpivirine, a second generation non-nucleoside reverse transcriptase inhibitors of HIV and dolutegravir, a novel integrase stand transfer inhibitors of HIV concentrations has been developed. Sample pre-treatment is limited to protein precipitation with a mixture of methanol and zinc sulfate. After centrifugation the supernatant is injected in the chromatographic system, which consists of on-line solid phase extraction followed by separation on a phenyl-hexyl column. This 2.5min method, with its simple sample preparation provides sensitive (the limit of quantitation is 25ng/mL for each compound), accurate and precise (the intra-day and inter-day imprecision and inaccuracy are lower than 15%) quantification of the plasma concentration of these drugs and can be used for therapeutic drug monitoring in patients infected with HIV.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%