  • Search Results
  • Pharmacological basis for the medicinal use of Linum usitatissimum (Flaxseed) in infectious and non-infectious diarrhea.
Pharmacological basis for the medicinal use of Linum usitatissimum (Flaxseed) in infectious and non-infectious diarrhea.
Journal of ethnopharmacology (2014-11-30)
Amber Hanif Palla, Naveed Ahmed Khan, Samra Bashir, Najeeb Ur-Rehman, Junaid Iqbal, Anwarul-Hassan Gilani
ABSTRACT
Linum usitatissimum, commonly known as Flaxseed has traditionally been used for the management of diarrhea and gastrointestinal infections. This study was planned to assess pharmacological basis for the medicinal use of Flaxseed in infectious and non-infectious diarrhea. The crude aqueous-methanolic extract of Flaxseed was studied using the in vivo castor oil-induced diarrhea, gut motility and enteropooling assays. Mechanistic basis was further elucidated by testing the inhibitory effect on spontaneously contracting isolated rabbit jejunum preparations, suspended in a 10ml tissue bath containing Tyrode׳ solution, maintained at 37°C and aerated with carbogen. Antibacterial efficacy of the Flaxseed extract was tested against different enteric and non-enteric pathogenic bacteria using in vitro antibacterial assays. Flaxseed extract reduced the diarrheal score in mice, by 39%, 63.90% and 68.34% at the respective doses of 100, 300 and 500mg/kg. Intestinal secretions were reduced by 24.12%, 28.09% and 38.80%, whereas the intestinal motility was reduced by 31.66%, 46.98% and 56.20% at respective doses of 100, 300 and 500mg/kg. When tested on isolated rabbit jejunum preparations, Flaxseed extract produced a dose-dependent inhibition of both spontaneous and high K(+) (80mM)-induced contractions, and shifted the concentration-response curves of Ca(++) to the right with suppression of the maximum response, similar to that caused by verapamil. Flaxseed extract was found to possess bactericidal activity at the tested concentrations of 12.5mg/ml, against vancomycin-resistant Enterococcus faecalis (100%), Escherichia coli K1 (88.88%), methicillin-resistant Staphylococcus aureus (98.76%), Bacillus cereus (92.64%), Pseudomonas aeruginosa (76.83%) and Salmonella typhi (26.91±3.35%). The concentration of 10mg/ml showed bactericidal effects against all the aforementioned pathogens except Escherichia coli K1, whereas for Pseudomonas aeruginosa and Salmonella typhi, it was bacteriostatic at this concentration. Our results indicate that Linum usitatissimum (Flaxseed) extract exhibits antidiarrheal and antispasmodic activities by virtue of its antimotility and antisecretory effects which are mediated possibly through inhibition of Ca(++) channels, though additional mechanism(s) cannot be ruled out. Flaxseed extract proved effective against both enteric and non-enteric pathogens causing diarrhea, thus ensuring wide coverage and rationalizing its medicinal use in both the infectious and non-infectious diarrhea.
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium phosphate monobasic, 99.998% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma