  • Search Results
  • Curcumin and quercetin trigger apoptosis during benzo(a)pyrene-induced lung carcinogenesis.
Curcumin and quercetin trigger apoptosis during benzo(a)pyrene-induced lung carcinogenesis.
Molecular and cellular biochemistry (2014-11-02)
Praveen Nair, Anshoo Malhotra, Devinder K Dhawan
ABSTRACT
The aim of the present study was to assess the cumulative effects of curcumin and quercetin in inducing apoptosis during benzo(a)pyrene (BP) (100 mg/Kg body weight)-induced lung carcinogenesis in mice. BP treatment resulted in a significant increase in the protein expression of Bcl-2 whereas expression of Bax was significantly decreased. Further, BP treatment brought about a significant decrease in the activities of caspase 3, caspase 9 as well as the number of apoptotic cells. Interestingly, separate as well as combined supplementation of curcumin (60 mg/kg body weight) and quercetin (40 mg/kg body weight) to BP-treated animals resulted in a significant decrease in the protein expression of Bcl-2 but caused a significant increase in the protein expression of Bax along with a noticeable improvement in the number of apoptotic cells. Also, supplementation with curcumin and quercetin separately to BP-treated mice brought a significant improvement in the enzyme activities of caspase 9 as well as caspase 3 but the improvement was more pronounced following combined treatment. Therefore, curcumin and quercetin, if given in combination shall exhibit enhanced chemopreventive potential against development of lung carcinogenesis by stimulating the apoptotic machinery.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Curcumin, analytical standard
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Curcumin, primary reference standard