Merck
  • Hawthorne effect with transient behavioral and biochemical changes in a randomized controlled sleep extension trial of chronically short-sleeping obese adults: implications for the design and interpretation of clinical studies.
Hawthorne effect with transient behavioral and biochemical changes in a randomized controlled sleep extension trial of chronically short-sleeping obese adults: implications for the design and interpretation of clinical studies.
PloS one (2014-08-21)
Giovanni Cizza, Paolo Piaggi, Kristina I Rother, Gyorgy Csako
ABSTRACT
To evaluate the effects of study participation per se at the beginning of a sleep extension trial between screening, randomization, and the run-in visit. Subjects were screened, returned for randomization (Comparison vs. Intervention) after 81 days (median), and attended run-in visit 121 days later. Outpatient. Obese (N = 125; M/F, 30/95; Blacks/Whites/Other, N = 73/44/8), mean weight 107.6±19.7 kg, <6.5 h sleep/night. Non-pharmacological sleep extension. Sleep duration (diaries and actigraphy watch), sleep quality (Pittsburgh Sleep Quality Index), daily sleepiness (Epworth Sleepiness Scale), fasting glucose, insulin and lipids. Prior to any intervention, marked improvements occurred between screening and randomization. Sleep duration increased (diaries: 357.4 ±51.2 vs. 388.1±48.6 min/night; mean±SD; P<0.001 screening vs. randomization; actigraphy: 344.3 ±41.9 vs. 358.6±48.2 min/night; P<0.001) sleep quality improved (9.1±3.2 vs. 8.2±3.0 PSQI score; P<0.001), sleepiness tended to improve (8.9±4.6 vs. 8.3±4.5 ESS score; P = 0.06), insulin resistance decreased (0.327±0.038 vs. 0.351±0.045; Quicki index; P<0.001), and lipids improved, except for HDL-C. Abnormal fasting glucose (25% vs. 11%; P = 0.007), and metabolic syndrome (42% vs. 29%; P = 0.007) both decreased. In absence of intervention, the earlier metabolic improvements disappeared at the run-in visit. Relatively small sample size. Improvements in biochemical and behavioral parameters between screening and randomization changed the "true" study baseline, thereby potentially affecting outcome. While regression to the mean and placebo effect were considered, these findings are most consistent with the "Hawthorne effect", according to which behavior measured in the setting of an experimental study changes in response to the attention received from study investigators. This is the first time that biochemical changes were documented with respect to the Hawthorne effect. The findings have implications for the design and conduct of clinical research. ClinicalTrials.gov NCT00261898.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material