  • Search Results
  • Modulation of RTK by sEcad: a putative mechanism for oncogenicity in oropharyngeal SCCs.
Modulation of RTK by sEcad: a putative mechanism for oncogenicity in oropharyngeal SCCs.
Oral diseases (2014-03-13)
X Teng, L Ma, S Kyrkanides, V Raja, D Trochesset, S M Brouxhon
ABSTRACT
Heightened levels of sEcad are found in the serum of patients with cancer and correlate with an unfavorable prognosis and later-stages of disease. In this study, we explored whether sEcad is elevated in human OPSCC specimens and FaDu cells. Additionally, we investigated sEcad-EGFR and sEcad-IGF-1R interactions and performed a functional analysis of sEcad in OPSCC cancers. sEcad, EGFR, and IGF-1R levels were examined in human OPSCC specimens and cells by immunoblotting. sEcad-EGFR and sEcad-IGF-1R interactions were examined by immunoprecipitation and immunoblot assays. Levels of sEcad on EGFR and IGF-1R pathway components were evaluated by IB. The effects of sEcad on OPSCC proliferation, migration, and invasion were assessed using standard cellular assays. Statistical analysis demonstrated that sEcad levels were significantly higher in OPSCC primary tumors and cells compared with normal controls. IP studies indicated that sEcad associated with EGFR and IGF-1R, and addition of sEcad resulted in a statistically significant increase in downstream signaling. Finally, cell-based assays demonstrated enhanced sEcad-induced proliferation, migration, and invasion, which was blocked by EGFR and IGF-1R inhibitors. These findings suggest that sEcad may play an important role in OPSCC oncogenicity via its interaction and activation of EGFR and IGF-1R.
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerol, analytical standard
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)