  • Search Results
  • Antinociceptive and antidiarrheal effects of pioglitazone in a rat model of diarrhoea-predominant irritable bowel syndrome: role of nitric oxide.
Antinociceptive and antidiarrheal effects of pioglitazone in a rat model of diarrhoea-predominant irritable bowel syndrome: role of nitric oxide.
Clinical and experimental pharmacology & physiology (2014-01-30)
Pedram Paragomi, Reza Rahimian, Mohammad Hossein Kazemi, Mohammad Hadi Gharedaghi, Amin Khalifeh-Soltani, Saeedeh Azary, Abbas Norouzi Javidan, Kamran Moradi, Stephen Sakuma, Ahmad Reza Dehpour
ABSTRACT
Irritable bowel syndrome (IBS) is a prevalent disease characterized by abdominal pain and abnormal bowel habits. Pioglitazone is a peroxisome proliferator-activated receptor (PPAR) γ agonist and, although it is mostly used as an antidiabetic agent, it has been reported to have analgesic effects. Nitric oxide (NO), a gaseous molecule that mediates many of the effects of pioglitazone, has been implicated in the pathophysiology of IBS. The aim of the present study was to investigate the effects of pioglitazone on symptoms in a rat model of diarrhoea-predominant IBS (D-IBS).and to determine the role of NO in these effects. Diarrhoea-predominant IBS was induced by intracolonic instillation of acetic acid. Pioglitazone (2 mg/kg, i.p.) was administered on Days 7, 9 and 11 after acetic acid instillation. To investigate the mechanism involved in pioglitazone action, rats were also administered either the PPARγ antagonist GW9662 (3 mg/kg, i.p.), the NO synthase (NOS) inhibitor N(G) -nitro-l-arginine methyl ester (l-NAME; 10 mg/kg, i.p.) or the NO precursor l-arginine (250 mg/kg, i.p.) along with pioglitazone. Visceral hypersensitivity, nociceptive thresholds, defecation frequency, stool form, serum and colon NO production and inducible (i) NOS activity were assessed 1 h after the final injection of pioglitazone or dimethylsulphoxide (used as the vehicle). Pioglitazone reduced visceral hypersensitivity and defecation frequency, increased nociceptive thresholds, NO production and iNOS activity and shifted stool form towards hard stools in D-IBS rats. These effects of pioglitazone were significantly reversed by l-NAME, but not GW9662. l-Arginine augmented the effects of pioglitazone. In conclusion, pioglitazone alleviates symptoms in a rat model of D-IBS through an NO-dependent mechanism.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
2-Chloro-5-nitrobenzanilide, 97%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
GW9662, >98% (HPLC)
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard