  • Search Results
  • Orexin-1 receptor antagonist in central nucleus of the amygdala attenuates the acquisition of flavor-taste preference in rats.
Orexin-1 receptor antagonist in central nucleus of the amygdala attenuates the acquisition of flavor-taste preference in rats.
Pharmacology, biochemistry, and behavior (2014-09-17)
Severiano Risco, Cristina Mediavilla
ABSTRACT
Previous studies demonstrated that the intracerebroventricular administration of SB-334867-A, a selective antagonist of orexin OX1R receptors, blocks the acquisition of saccharin-induced conditioned flavor preference (CFP) but not LiCl-induced taste aversion learning (TAL). Orexinergic fibers from the lateral hypothalamus end in the central nucleus of the amygdala (CeA), which expresses orexin OX1R receptors. Taste and sensory inputs also are present in CeA, which may contribute to the development of taste learning. This study analyzed the effect of two doses (1.5 and 6μg/0.5μl) of SB-334867-A administered into the CeA on flavor-taste preference induced by saccharin and on TAL induced by a single administration of LiCl (0.15M, 20ml/kg, i.p.). Outcomes indicate that inactivation of orexinergic receptors in the CeA attenuates flavor-taste preference in a two-bottle test (saccharin vs. water). Intra-amygdalar SB-334867-A does not affect gustatory processing or the preference for the sweet taste of saccharin given that SB-334867-A- and DMSO-treated groups (control animals) increased the intake of the saccharin-associated flavor across training acquisition sessions. Furthermore, SB-334867-A in the CeA does not block TAL acquisition ruling out the possibility that functional inactivation of OX1R receptors interferes with taste processing. Orexin receptors in the CeA appear to intervene in the association of a flavor with orosensory stimuli, e.g., a sweet and pleasant taste, but could be unnecessary when the association is established with visceral stimuli, e.g., lithium chloride. These data suggest that orexinergic projections to the CeA may contribute to the reinforcing signals facilitating the acquisition of taste learning and the change in hedonic evaluation of the taste, which would have important implications for the OX1R-targeted pharmacological treatment of eating disorders.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%