  • Search Results
  • Inhibitory action of pristimerin on hypoxia‑mediated metastasis involves stem cell characteristics and EMT in PC-3 prostate cancer cells.
Inhibitory action of pristimerin on hypoxia‑mediated metastasis involves stem cell characteristics and EMT in PC-3 prostate cancer cells.
Oncology reports (2015-01-13)
Jianwei Zuo, Yuanqing Guo, Xinsheng Peng, Yubo Tang, Xintao Zhang, Peiheng He, Shuaihua Li, Qingde Wa, Jinglei Li, Shuai Huang, Dongliang Xu
ABSTRACT
The aim of the present study was to investigate whether pristimerin affects the bone metastasis, stem cell characteristics and epithelial-mesenchymal transition (EMT) of prostate cancer (PCa) PC-3 cells subjected to hypoxia. The PC-3 cells were cultured under hypoxia or normoxia for 48 h and were then treated with increasing concentrations of pristimerin from 0 to 0.8 µmol/l, under normoxia. Hypoxia‑inducible factor-1α (HIF-1α) was detected by western blotting. Proliferation was assessed with the CCK-8 assay. Transwell invasion assay was used to analyze the potency of invasion. Stem cell characteristics were detected by sphere formation, colony formation assay and western blotting, including CD44, KLF4, OCT4 and AGO2, which are stem cell characteristic-related markers. EMT was confirmed by the expression changes of EMT-related markers, including N-cadherin, fibronectin, vimentin and ZEB1, which were evaluated by western blotting. The addition of pristimerin to the medium reduced the hypoxia-induced PC-3 cell proliferation in a dose-dependent manner. Pristimerin effectively inhibited hypoxia‑induced invasion of the PCa cells in vitro. Moreover, the treatment of cells with pristimerin induced the reversal of hypoxia-induced stem cell characteristics and EMT, which was confirmed by sphere formation, colony formation assay and the expression changes of CSC- and EMT-related markers. The reversal of hypoxia‑induced stem cell characteristics and EMT in the PCa cells by low-dose pristimerin was dose‑dependent. These results showed that treatment with pristimerin may be a potential strategy for the suppression of hypoxia-induced metastasis through the reversal of hypoxia-induced stem cell characteristics and EMT in cancer cells, which justifies the potential use of pristimerin as a practical chemopreventive approach for patients with PCa.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)