  • Search Results
  • Rosiglitzone suppresses angiotensin II-induced production of KLF5 and cell proliferation in rat vascular smooth muscle cells.
Rosiglitzone suppresses angiotensin II-induced production of KLF5 and cell proliferation in rat vascular smooth muscle cells.
PloS one (2015-04-16)
Dengfeng Gao, Guanghua Hao, Zhe Meng, Ning Ning, Guang Yang, Zhongwei Liu, Xin Dong, Xiaolin Niu
ABSTRACT
Krüppel-like factor (KLF) 5, which initiates vascular smooth muscle cell (VSMC) proliferation, also participates in Angiotensin (Ang) II-induced vascular remodeling. The protective effect of rosiglitazone on vascular remodeling may be due to their impact on VSMC proliferation. However, the underlying mechanisms involved remain unclear. This study was designed to investigate whether the antiproliferation effects of rosiglitazone are mediated by regulating Ang II/KLF5 response. We found that, in aortas of Ang II-infused rats, vascular remodeling and KLF5 expression were markedly increased, and its target gene cyclin D1 was overexpressed. Co-treatment with rosiglitazone diminished these changes. In growth-arrested VSMCs, PPAR-γ agonists (rosiglitazone and 15d-PGJ2) dose-dependently inhibited Ang II-induced cell proliferation and expression of KLF5 and cyclin D1. Moreover, these effects were attenuated by the PPAR-γ antagonists GW9662, bisphenol A diglycidyl ether and PPAR-γ specific siRNA. Furthermore, rosiglitazone inhibited Ang II-induced phosphorylation of protein kinase C (PKC) ζ and extracellular signal-regulated kinase (ERK) 1/2 and activation of early growth response protein (Egr). In conclusion, in Ang II-stimulated VSMCs, rosiglitazone might have an antiproliferative effect through mechanisms that include reducing KLF5 expression, and a crosstalk between PPAR-γ and PKCζ/ERK1/2/Egr may be involved in. These findings not only provide a previously unrecognized mechanism by which PPAR-γ agonists inhibit VSMC proliferation, but also document a novel evidence for the beneficial vascular effect of PPAR-γ activation.
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
2-Chloro-5-nitrobenzanilide, 97%
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
GW9662, >98% (HPLC)