  • Search Results
  • Early hematological and immunological alterations in gasoline station attendants exposed to benzene.
Early hematological and immunological alterations in gasoline station attendants exposed to benzene.
Environmental research (2015-01-21)
Angela M Moro, Natália Brucker, Mariele F Charão, Elisa Sauer, Fernando Freitas, Juliano Durgante, Guilherme Bubols, Sarah Campanharo, Rafael Linden, Ana P Souza, Cristina Bonorino, Rafael Moresco, Diogo Pilger, Adriana Gioda, Sandra Farsky, Albert Duschl, Solange C Garcia
ABSTRACT
Elucidation of effective biomarkers may provide tools for the early detection of biological alterations caused by benzene exposure and may contribute to the reduction of occupational diseases. This study aimed to assess early alterations on hematological and immunological systems of workers exposed to benzene. Sixty gasoline station attendants (GSA group) and 28 control subjects were evaluated. Environmental and biological monitoring of benzene exposure was performed in blood and urine. The potential effect biomarkers evaluated were δ-aminolevulinate dehydratase (ALA-D) activity, CD80 and CD86 expression in lymphocytes and monocytes, and serum interleukin-8 (IL-8). The influence of confounding factors and toluene co-exposure were considered. Although exposures were below ACGIH (American Conference of Governmental Industrial Hygienists) limits, reduced ALA-D activity, decreased CD80 and CD86 expression in monocytes and increased IL-8 levels were found in the GSA group compared to the control subjects. Furthermore, according to multiple linear regression analysis, benzene exposure was associated to a decrease in CD80 and CD86 expression in monocytes. These findings suggest, for the first time, a potential effect of benzene exposure on ALA-D activity, CD80 and CD86 expression, IL-8 levels, which could be suggested as potential markers for the early detection of benzene-induced alterations.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Toluene, analytical standard
Supelco
Dichloromethane, analytical standard
Supelco
Benzene, analytical standard
Supelco
p-Xylene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)