  • Search Results
  • The thymoquinone-induced production of reactive oxygen species promotes dedifferentiation through the ERK pathway and inflammation through the p38 and PI3K pathways in rabbit articular chondrocytes.
The thymoquinone-induced production of reactive oxygen species promotes dedifferentiation through the ERK pathway and inflammation through the p38 and PI3K pathways in rabbit articular chondrocytes.
International journal of molecular medicine (2014-12-02)
Seon-Mi Yu, Song-Ja Kim
ABSTRACT
Dedifferentiation and inflammation are major features of cartilage degeneration during the pathogenesis of osteoarthritis (OA). Thymoquinone (TQ) is the major compound of black seed oil isolated from Nigella sativa with various beneficial or harmful effects on several diseases; however, its effects on the dedifferentiation and inflammation of chondrocytes have not yet been characterized. In the present study, we investigated whether TQ regulates the dedifferentiation and inflammation of rabbit articular chondrocytes, focusing on the production of reactive oxygen species (ROS) in rabbit articular chondrocytes. TQ induced the generation of ROS in a dose-dependent manner, as shown by staining with the fluorescent probe, 2'-7'-dichlorofluorescein diacetate. We confirmed that TQ induced dedifferentiation by measuring the loss of type II collagen and the reduction in chondroitin sulfate proteoglycan levels. TQ also caused inflammation by inducing the expression of cyclooxygenase-2 (COX-2) and prostaglandin E2 (PGE2). The antioxidant, N-acetyl cysteine (NAC), prevented the dedifferentiation and inflammation which was generated by the TQ-induced production of ROS. Furthermore, TQ caused a dose-dependent increase in p38, phosphorylated extracellular signal-regulated kinase (p-ERK) and phosphoinositide 3-kinase (PI3K) expression. NAC abrogated this effect and attenuated the dedifferentiation and inflammation which was generated by the TQ-induced production of ROS. To identify the ROS-regulated pathways, we treated the chondrocytes with the p38 inhibitor, SB203580, the MEK inhibitor, PD98059, and the PI3K inhibitor, LY294002. PD98059 inhibited the TQ-induced dedifferentiation and SB203580 and LY294002 prevented the TQ-induced inflammation. These findings suggest that the TQ-induced production of ROS causes dedifferentiation through the ERK pathway and inflammation through the PI3K and p38 pathways in rabbit articular chondrocytes.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
4,4′-Diisothiocyanatostilbene-2,2′-disulfonic acid disodium salt hydrate, ≥80% (elemental analysis), powder
Supelco
Dimethyl sulfoxide, analytical standard