  • Search Results
  • Selective analysis of human serum albumin based on SEC-ICP-MS after labelling with iophenoxic acid.
Selective analysis of human serum albumin based on SEC-ICP-MS after labelling with iophenoxic acid.
Analytical and bioanalytical chemistry (2015-02-05)
Julie Maria Dersch, Tam T T N Nguyen, Jesper Østergaard, Stefan Stürup, Bente Gammelgaard
ABSTRACT
Human serum albumin (HSA) is the most abundant protein in the human plasma. HSA has several physiological roles in the human body, including storage and transport. Owing to the predominance of albumin in plasma, HSA is often involved in the protein binding of drugs. The aim of this work was to develop a selective, quantitative method for determining albumin in plasma with the purpose of clarifying the fate of metal-based drugs in biological systems. The method can also be applied for determination of urine albumin, which is of relevance in diagnostics of kidney disease. A selective method for quantification of HSA based on labelling the protein with iophenoxic acid (IPA) was developed. Samples were subjected to size exclusion chromatography (SEC) and detection by inductively coupled plasma mass spectrometry (ICP-MS) monitoring iodine and platinum. The iodine signal for the HSA-IPA complex showed linearity in the range 1 to 250 mg L(-1). The precision was 3.7% and the accuracy 100.7% determined by analysis of a certified HSA reference material. The limit of detection (LOD) and limit of quantification (LOQ) were 0.23 and 9.79 mg L(-1), respectively. The method was applied for analysis of HSA in human plasma and urine samples and for studying the binding of cisplatin to proteins in the human plasma.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
α-Ethyl-3-hydroxy-2,4,6-triiodohydrocinnamic acid, 97%
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC