Merck
  • Discovery and preclinical studies of (R)-1-(4-(4-fluoro-2-methyl-1H-indol-5-yloxy)-5- methylpyrrolo[2,1-f][1,2,4]triazin-6-yloxy)propan- 2-ol (BMS-540215), an in vivo active potent VEGFR-2 inhibitor.
Discovery and preclinical studies of (R)-1-(4-(4-fluoro-2-methyl-1H-indol-5-yloxy)-5- methylpyrrolo[2,1-f][1,2,4]triazin-6-yloxy)propan- 2-ol (BMS-540215), an in vivo active potent VEGFR-2 inhibitor.
Journal of medicinal chemistry (2006-03-31)
Rajeev S Bhide, Zhen-Wei Cai, Yong-Zheng Zhang, Ligang Qian, Donna Wei, Stephanie Barbosa, Louis J Lombardo, Robert M Borzilleri, Xiaoping Zheng, Laurence I Wu, Joel C Barrish, Soong-Hoon Kim, Kenneth Leavitt, Arvind Mathur, Leslie Leith, Sam Chao, Barri Wautlet, Steven Mortillo, Robert Jeyaseelan, Daniel Kukral, John T Hunt, Amrita Kamath, Aberra Fura, Viral Vyas, Punit Marathe, Celia D'Arienzo, George Derbin, Joseph Fargnoli
ABSTRACT
A series of substituted 4-(4-fluoro-1H-indol-5-yloxy)pyrrolo[2,1-f][1,2,4]triazine-based inhibitors of vascular endothelial growth factor receptor-2 kinase is reported. Structure-activity relationship studies revealed that a methyl group at the 5-position and a substituted alkoxy group at the 6-position of the pyrrolo[2,1-f][1,2,4]triazine core gave potent compounds. Biochemical potency, kinase selectivity, and pharmacokinetics of the series were optimized and in vitro safety liabilities were minimized to afford BMS-540215 (12), which demonstrated robust preclinical in vivo activity in human tumor xenograft models. The l-alanine prodrug of 12, BMS-582664 (21), is currently under evaluation in clinical trials for the treatment of solid tumors.
Sigma-Aldrich
Brivanib, ≥98% (HPLC)