  • Search Results
  • Troxerutin, a plant flavonoid, protects cells against oxidative stress-induced cell death through radical scavenging mechanism.
Troxerutin, a plant flavonoid, protects cells against oxidative stress-induced cell death through radical scavenging mechanism.
Food chemistry (2015-10-17)
Niranjan A Panat, Dharmendra K Maurya, Saroj S Ghaskadbi, Santosh K Sandur
ABSTRACT
Troxerutin (TRX) is a flavonoid present in tea, coffee, cereal grains, various fruits and vegetables have been reported to exhibit radioprotective, antithrombotic, nephro and hepato-protective effects. A systematic study was undertaken to evaluate its free radical scavenging ability and anti-apoptotic activity in cell-free and cellular systems. TRX scavenged superoxide, nitric oxide and also other model stable radicals like 1,1-diphenyl-2-picryl-hydazyl, and 2,2'-azinobis3-ethylbenzothiazoline-6-sulfonic acid. It reacted with hydroxyl radicals, carbonate and thiocyanate anions, as evaluated by pulse radiolysis and stopped flow techniques. TRX protected different cell types (epithelial cells, fibroblasts and lymphocytes) against peroxyl radical-induced apoptosis, necrosis and mitotic death. It scavenged intracellular basal and inducible ROS levels and also restored depletion of intracellular GSH levels, suggesting that free radical scavenging ability may be responsible for the observed cytoprotection of different cell types. TRX may find application as an adjuvant in treating various diseases attributed to oxidative stress.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Phenazine methosulfate, ≥90% (UV)
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
JC-1, solid
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh