  • Search Results
  • Melatonin exerts protective effect on N2a cells under hypoxia conditions through Zip1/ERK pathway.
Melatonin exerts protective effect on N2a cells under hypoxia conditions through Zip1/ERK pathway.
Neuroscience letters (2015-04-14)
Xin-Wei Liu, Ying Zi, Yun-En Liu, Yu-Biao Zhang, Liang-Bi Xiang, Ming-xiao Hou
ABSTRACT
Melatonin plays a neuroprotective role in different CNS injuries. However, the molecular mechanisms underlying neuroprotection by melatonin are not well understood. Here, we studied the effects of melatonin in hypoxia-induced N2a cells and our results demonstrated that melatonin not only reduced the level of ROS and MDA, induced the increase of SOD, but also increased the cell proliferation and inhibited cell apoptosis in hypoxia-induced N2a cells. Moreover, we identified that melatonin can activate the MAPK/ERK pathway via upregulating the expression of Zip1. Therefore, this study provides a new mechanism of melatonin and need our further study in detail.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Fluorescein, for fluorescence, free acid